Assay Details
Target Gene Details
Entrez Gene ID: | 10061 |
Gene Name: | ATP binding cassette subfamily F member 2 |
Gene Aliases: |
ABC28, EST133090, HUSSY-18, HUSSY18 |
Location: |
Chr.7:151207837-151227374 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCF2 | NM_005692.4 | NP_005683.2 | ||
| AL050291.1 | CAB43392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609035 | Chr.7:151199137 - 151258679 on Build GRCh38 | Loss |
|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113812349] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map