Assay Details
Target Gene Details
Entrez Gene ID: | 7589 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
NY-REN-21, ZNF38, Zipro1 |
Location: |
Chr.7:100049770-100065041 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN21 | NM_145914.2 | NP_666019.1 | ||
| XM_005250568.4 | XP_005250625.1 | |||
| XM_006716113.3 | XP_006716176.1 | |||
| XM_017012584.1 | XP_016868073.1 | |||
| XM_017012585.1 | XP_016868074.1 | |||
| AF155100.1 | AAD42866.1 | |||
| AK075324.1 | ||||
| AK292267.1 | ||||
| AK301871.1 | ||||
| AK313884.1 | ||||
| AL136865.1 | CAB66799.1 | |||
| BC047309.1 | AAH47309.1 | |||
| BI764964.1 | ||||
| CR749301.1 | CAH18156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map