Assay Details
Target Gene Details
Entrez Gene ID: | 51606 |
Gene Name: | ATPase H+ transporting V1 subunit H |
Gene Aliases: |
CGI-11, MSTP042, NBP1, SFD, SFDalpha, SFDbeta, VMA13 |
Location: |
Chr.8:53715543-53843311 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1H | NM_015941.3 | NP_057025.2 | ||
| NM_213619.2 | NP_998784.1 | |||
| NM_213620.2 | NP_998785.1 | |||
| XM_006716455.2 | XP_006716518.1 | |||
| XM_011517542.1 | XP_011515844.1 | |||
| AF112204.1 | AAF17192.1 | |||
| AF113222.1 | AAG39293.1 | |||
| AF132945.1 | AAD27720.1 | |||
| AF298777.1 | AAG22809.1 | |||
| AI741756.1 | ||||
| AK022345.1 | ||||
| AK094839.1 | ||||
| AK098305.1 | ||||
| AK303229.1 | ||||
| AK308204.1 | ||||
| BC025275.1 | AAH25275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759616 | Chr.8:53562029 - 53824913 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73682552] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map