Assay Details
Target Gene Details
Entrez Gene ID: | 619373 |
Gene Name: | membrane bound O-acyltransferase domain containing 4 |
Gene Aliases: |
FKSG89, GOAT, OACT4 |
Location: |
Chr.8:30131454-30148768 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBOAT4 | NM_001100916.1 | NP_001094386.1 | ||
| XM_017013726.1 | XP_016869215.1 | |||
| EU518495.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831277 | Chr.8:29977428 - 30180297 on Build GRCh38 | Gain |
|
| esv3616784 | Chr.8:30138142 - 30146624 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map