Assay Details
Target Gene Details
Entrez Gene ID: | 100507530 |
Gene Name: | MCPH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:6615603-6708209 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCPH1-AS1 | NR_125386.1 | |||
| BX374425.2 | ||||
| DA281269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021684 | Chr.8:6492570 - 6951933 on Build GRCh38 | Loss |
|
| nsv610004 | Chr.8:6544845 - 6915391 on Build GRCh38 | Gain |
|
| nsv1025976 | Chr.8:6615427 - 6829450 on Build GRCh38 | Loss |
|
| nsv524670 | Chr.8:6643023 - 6682466 on Build GRCh38 | Loss |
|
| esv2422383 | Chr.8:6504868 - 6924071 on Build GRCh38 | Duplication |
|
| nsv610005 | Chr.8:6633144 - 6678169 on Build GRCh38 | Gain |
|

Genomic Map