Assay Details
Target Gene Details
Entrez Gene ID: | 100130742 |
Gene Name: | leucine rich repeat containing 69 |
Gene Aliases: |
- |
Location: |
Chr.8:91102619-91219236 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC69 | NM_001129890.1 | NP_001123362.1 | ||
| AK097393.1 | ||||
| AK130865.1 | BAC85448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2656822 | Chr.8:91103277 - 91173399 on Build GRCh38 | Deletion |
|
| esv3572901 | Chr.8:91101144 - 91173507 on Build GRCh38 | Loss |
|
| nsv611721 | Chr.8:91094842 - 91171430 on Build GRCh38 | Loss |
|
| nsv831390 | Chr.8:91024783 - 91186377 on Build GRCh38 | Gain |
|
| nsv465736 | Chr.8:91110980 - 91161887 on Build GRCh38 | Loss |
|
| esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss |
|
| dgv7268n100 | Chr.8:91106767 - 91173829 on Build GRCh38 | Loss |
|
| esv3618014 | Chr.8:91103260 - 91173374 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map