Assay Details
Target Gene Details
Entrez Gene ID: | 101927245 |
Gene Name: | vegetative cell wall protein gp1-like |
Gene Aliases: |
- |
Location: |
Chr.8:102654313-102730620 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927245 | XM_011517419.2 | XP_011515721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|

Genomic Map