Assay Details
Target Gene Details
Entrez Gene ID: | 157773 |
Gene Name: | chromosome 8 open reading frame 48 |
Gene Aliases: |
- |
Location: |
Chr.8:13566843-13568288 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf48 | NM_001007090.2 | 1 | 869 | NP_001007091.1 |
| AF394908.1 | 1 | 722 | AAQ02889.1 | |
| AK058131.1 | 1 | 869 | BAB71677.1 | |
| AK058156.1 | 1 | 249 | BAB71692.1 | |
| BC031245.1 | 1 | 817 | AAH31245.1 | |
| CB306127.1 | 1 | 595 |
Target Gene Details
Entrez Gene ID: | 10395 |
Gene Name: | DLC1 Rho GTPase activating protein |
Gene Aliases: |
ARHGAP7, HP, STARD12, p122-RhoGAP |
Location: |
Chr.8:13083361-13604606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLC1 | XM_017012951.1 | XP_016868440.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517019 | Chr.8:13511552 - 14801106 on Build GRCh38 | Gain+Loss |
|
| esv3616315 | Chr.8:13567359 - 13968374 on Build GRCh38 | Loss |
|
| nsv610412 | Chr.8:13121891 - 13662651 on Build GRCh38 | Gain |
|
| nsv8290 | Chr.8:13234049 - 13677278 on Build GRCh38 | Gain |
|
| nsv1032823 | Chr.8:13472267 - 13762903 on Build GRCh38 | Gain |
|
| nsv1034745 | Chr.8:13524571 - 13670170 on Build GRCh38 | Gain |
|
| nsv610411 | Chr.8:13093516 - 14267300 on Build GRCh38 | Loss |
|
| nsv1022834 | Chr.8:13067043 - 14121780 on Build GRCh38 | Gain |
|
| nsv610420 | Chr.8:13469408 - 13765962 on Build GRCh38 | Gain |
|
| nsv470185 | Chr.8:13539114 - 13573069 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map