Assay Details
Target Gene Details
Entrez Gene ID: | 389643 |
Gene Name: | nuclear GTPase, germinal center associated |
Gene Aliases: |
C8orf80, HMFN0672, SLIP-GC |
Location: |
Chr.8:28021964-28084068 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUGGC | NM_001010906.1 | NP_001010906.1 | ||
| XM_011544523.2 | XP_011542825.1 | |||
| XM_011544524.2 | XP_011542826.1 | |||
| XM_011544525.1 | XP_011542827.1 | |||
| AA482187.1 | ||||
| AB075870.1 | BAD38652.1 | |||
| AK129923.1 | BAC85251.1 |
More Information
Set Membership: |
|

Genomic Map