Assay Details
Target Gene Details
Entrez Gene ID: | 10863 |
Gene Name: | ADAM metallopeptidase domain 28 |
Gene Aliases: |
ADAM 28, MDC-L, MDCL, eMDC II, eMDCII |
Location: |
Chr.8:24294040-24359018 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM28 | NM_001304351.1 | NP_001291280.1 | ||
| NM_014265.5 | NP_055080.2 | |||
| NM_021777.4 | NP_068547.2 | |||
| NR_130709.1 | ||||
| NR_130710.1 | ||||
| XM_005273380.3 | XP_005273437.1 | |||
| XM_005273382.1 | XP_005273439.1 | |||
| XM_006716273.3 | XP_006716336.1 | |||
| XM_006716274.1 | XP_006716337.1 | |||
| XM_011544367.2 | XP_011542669.1 | |||
| XM_011544368.2 | XP_011542670.1 | |||
| XM_011544369.2 | XP_011542671.1 | |||
| XM_011544370.2 | XP_011542672.1 | |||
| XM_017012974.1 | XP_016868463.1 | |||
| XM_017012975.1 | XP_016868464.1 | |||
| XM_017012976.1 | XP_016868465.1 | |||
| AF137334.1 | AAD25099.1 | |||
| AF137335.1 | AAD25100.1 | |||
| AJ242015.1 | CAB42085.1 | |||
| AK293382.1 | ||||
| AK303437.1 | ||||
| BC136478.1 | ||||
| DA933009.1 |
Target Gene Details
Entrez Gene ID: | 101929294 |
Gene Name: | uncharacterized LOC101929294 |
Gene Aliases: |
- |
Location: |
Chr.8:24295814-24548618 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929294 | NR_125808.1 | |||
| CD705133.1 | ||||
| DA221074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891377 | Chr.8:23722497 - 25043041 on Build GRCh38 | Loss |
|
| esv3616651 | Chr.8:24308855 - 24312163 on Build GRCh38 | Loss |
|
| nsv831271 | Chr.8:24203242 - 24391933 on Build GRCh38 | Gain |
|
| nsv610781 | Chr.8:24188182 - 24364512 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map