Assay Details
Target Gene Details
Entrez Gene ID: | 8601 |
Gene Name: | regulator of G-protein signaling 20 |
Gene Aliases: |
RGSZ1, ZGAP1, g(z)GAP, gz-GAP |
Location: |
Chr.8:53851808-53959304 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS20 | NM_001286673.1 | NP_001273602.1 | ||
| NM_001286674.1 | NP_001273603.1 | |||
| NM_001286675.1 | NP_001273604.1 | |||
| NM_170587.3 | NP_733466.1 | |||
| AF366054.1 | AAK54122.1 | |||
| AF366055.1 | AAK54123.1 | |||
| AF366056.1 | AAK54124.1 | |||
| AY046538.1 | AAL03971.1 |
More Information
Set Membership: |
|

Genomic Map