Assay Details
Target Gene Details
Entrez Gene ID: | 7336 |
Gene Name: | ubiquitin conjugating enzyme E2 V2 |
Gene Aliases: |
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2 |
Location: |
Chr.8:47997527-48063545 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2V2 | NM_003350.2 | NP_003341.1 | ||
| XM_011517583.2 | XP_011515885.1 | |||
| XM_017013808.1 | XP_016869297.1 | |||
| AF049140.1 | AAC05381.1 | |||
| AK313447.1 | ||||
| BC007051.1 | AAH07051.1 | |||
| BC016332.1 | AAH16332.1 | |||
| BC016710.1 | AAH16710.1 | |||
| BC028673.1 | AAH28673.1 | |||
| BC062418.1 | AAH62418.1 | |||
| BT006744.1 | AAP35390.1 | |||
| CR407628.1 | CAG28556.1 | |||
| U62136.2 | AAB04758.2 | |||
| X98091.1 | CAA66717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831309 | Chr.8:47902730 - 48065296 on Build GRCh38 | Loss |
|
| esv2759611 | Chr.8:47875732 - 48095454 on Build GRCh38 | Loss |
|
| nsv1032750 | Chr.8:47964619 - 48012268 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map