Assay Details
Target Gene Details
Entrez Gene ID: | 759 |
Gene Name: | carbonic anhydrase 1 |
Gene Aliases: |
CA-I, CAB, Car1, HEL-S-11 |
Location: |
Chr.8:85328229-85378154 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA1 | NM_001128829.3 | NP_001122301.1 | ||
| NM_001128830.3 | NP_001122302.1 | |||
| NM_001128831.3 | NP_001122303.1 | |||
| NM_001291967.1 | NP_001278896.1 | |||
| NM_001291968.1 | NP_001278897.1 | |||
| NM_001738.4 | NP_001729.1 | |||
| BC027890.1 | AAH27890.1 | |||
| BU655910.1 | ||||
| BU656154.1 | ||||
| BU658617.1 | ||||
| BU659301.1 | ||||
| BU659695.1 | ||||
| BU661390.1 | ||||
| BU663330.1 | ||||
| EU794595.1 | ||||
| M33987.1 | AAA51910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2658426 | Chr.8:85366147 - 85378751 on Build GRCh38 | Deletion |
|
| nsv831377 | Chr.8:85350878 - 85531912 on Build GRCh38 | Gain |
|
| dgv1406e214 | Chr.8:85366203 - 85379406 on Build GRCh38 | Loss |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map