Assay Details
Target Gene Details
Entrez Gene ID: | 9827 |
Gene Name: | RGP1 homolog, RAB6A GEF complex partner 1 |
Gene Aliases: |
KIAA0258 |
Location: |
Chr.9:35749280-35753267 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGP1 | NM_001080496.2 | NP_001073965.2 | ||
| AK289566.1 | ||||
| BC001725.1 | AAH01725.1 | |||
| D87447.1 |
More Information
Set Membership: |
|

Genomic Map