Assay Details
Target Gene Details
Entrez Gene ID: | 158038 |
Gene Name: | leucine rich repeat and Ig domain containing 2 |
Gene Aliases: |
LERN3, LRRN6C |
Location: |
Chr.9:27937617-29213981 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINGO2 | NM_001258282.1 | NP_001245211.1 | ||
| XM_011517719.1 | XP_011516021.1 | |||
| XM_017014303.1 | XP_016869792.1 | |||
| DA531255.1 |
Target Gene Details
Entrez Gene ID: | 100126310 |
Gene Name: | microRNA 876 |
Gene Aliases: |
MIRN876, hsa-mir-876, mir-876 |
Location: |
Chr.9:28863626-28863706 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR876 | NR_030597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471296 | Chr.9:28649449 - 28942610 on Build GRCh38 | Loss |
|
| dgv7498n100 | Chr.9:28847172 - 29183431 on Build GRCh38 | Loss |
|
| dgv12628n54 | Chr.9:28570654 - 28942610 on Build GRCh38 | Loss |
|
| nsv1020624 | Chr.9:28808326 - 29050315 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map