Assay Details
Target Gene Details
Entrez Gene ID: | 100289019 |
Gene Name: | SLC25A25 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:128111171-128118734 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A25-AS1 | NR_033374.1 | |||
| AK124723.1 | ||||
| AL833509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951769 | Chr.9:128101422 - 128129321 on Build GRCh38 | Deletion |
|
| nsv615383 | Chr.9:128091848 - 128128781 on Build GRCh38 | Loss |
|
| nsv818724 | Chr.9:128100434 - 128128781 on Build GRCh38 | Loss |
|
| nsv6723 | Chr.9:128106647 - 128141731 on Build GRCh38 | Deletion |
|
| nsv471324 | Chr.9:128039877 - 128130666 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75974900] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map