Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD8 | NM_001033113.1 | NP_001028285.1 | ||
| NM_198585.2 | NP_940987.2 | |||
| XM_011518665.2 | XP_011516967.1 | |||
| XM_011518666.2 | XP_011516968.1 | |||
| XM_011518668.1 | XP_011516970.1 | |||
| AY359088.1 | AAQ89446.1 | |||
| BC141810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss |
|
| nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion |
|
| nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss |
|
| esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss |
|
| nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114198953] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map