Assay Details
Target Gene Details
Entrez Gene ID: | 101927069 |
Gene Name: | uncharacterized LOC101927069 |
Gene Aliases: |
- |
Location: |
Chr.9:68953088-68975841 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927069 | NR_110647.1 | |||
| AK057188.1 |
Target Gene Details
Entrez Gene ID: | 8395 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 beta |
Gene Aliases: |
MSS4, STM7 |
Location: |
Chr.9:68705217-69009176 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP5K1B | NM_001278253.1 | NP_001265182.1 | ||
| NM_003558.3 | NP_003549.1 | |||
| XM_005252261.3 | XP_005252318.1 | |||
| XM_005252262.4 | XP_005252319.1 | |||
| XM_006717300.2 | XP_006717363.1 | |||
| XM_006717301.1 | XP_006717364.1 | |||
| XM_011519082.2 | XP_011517384.1 | |||
| XM_011519083.2 | XP_011517385.1 | |||
| XM_011519084.2 | XP_011517386.1 | |||
| XM_017015188.1 | XP_016870677.1 | |||
| XM_017015189.1 | XP_016870678.1 | |||
| XM_017015190.1 | XP_016870679.1 | |||
| XM_017015191.1 | XP_016870680.1 | |||
| AK292734.1 | ||||
| AK295587.1 | ||||
| BC030587.2 | AAH30587.1 | |||
| U78579.1 | AAC50914.1 | |||
| U78580.1 | AAC50915.1 | |||
| U78581.1 | AAC50916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891679 | Chr.9:68939243 - 69246082 on Build GRCh38 | Gain |
|
| dgv7666n100 | Chr.9:68896889 - 69228107 on Build GRCh38 | Gain |
|
| esv3576657 | Chr.9:68955164 - 69227475 on Build GRCh38 | Gain |
|
| esv2422258 | Chr.9:68619929 - 69048424 on Build GRCh38 | Duplication |
|
| nsv614548 | Chr.9:68856996 - 69320661 on Build GRCh38 | Gain |
|
| esv3891678 | Chr.9:68918451 - 69144626 on Build GRCh38 | Gain |
|
| nsv466393 | Chr.9:68838527 - 69089911 on Build GRCh38 | Gain |
|
| dgv7665n100 | Chr.9:68783797 - 69356369 on Build GRCh38 | Gain |
|
| nsv614550 | Chr.9:68959841 - 69228107 on Build GRCh38 | Gain |
|
| nsv614549 | Chr.9:68891543 - 69044364 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117542800] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map