Assay Details
Target Gene Details
Entrez Gene ID: | 347148 |
Gene Name: | pyroglutamylated RFamide peptide |
Gene Aliases: |
26RFa, P518 |
Location: |
Chr.9:130893428-130896812 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRFP | NM_198180.2 | 1 | 77 | NP_937823.1 |
| CV575364.1 | 1 | 77 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain |
|
| nsv951789 | Chr.9:130880714 - 130963813 on Build GRCh38 | Deletion |
|
| nsv469903 | Chr.9:130890269 - 130908179 on Build GRCh38 | Loss |
|
| nsv466601 | Chr.9:130894835 - 130934388 on Build GRCh38 | Loss |
|
| nsv831737 | Chr.9:130895103 - 131128557 on Build GRCh38 | Loss |
|
| esv3621869 | Chr.9:130876741 - 130992658 on Build GRCh38 | Loss |
|
| esv2759717 | Chr.9:130862179 - 131153174 on Build GRCh38 | Loss |
|
| nsv6735 | Chr.9:130877388 - 130921852 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79567040] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map