Assay Details
Target Gene Details
Entrez Gene ID: | 664 |
Gene Name: | BCL2 interacting protein 3 |
Gene Aliases: |
NIP3 |
Location: |
Chr.10:131967683-131982013 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIP3 | NM_004052.3 | NP_004043.3 | ||
| AF002697.1 | AAC16738.1 | |||
| AK222626.1 | BAD96346.1 | |||
| AK295136.1 | ||||
| BC021989.1 | AAH21989.1 | |||
| BC067818.1 | AAH67818.1 | |||
| BC080643.1 | AAH80643.1 | |||
| U15174.1 | AAC00022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv825650 | Chr.10:131914064 - 132056715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map