Assay Details
Target Gene Details
Entrez Gene ID: | 36 |
Gene Name: | acyl-CoA dehydrogenase, short/branched chain |
Gene Aliases: |
2-MEBCAD, ACAD7, SBCAD |
Location: |
Chr.10:123008913-123058290 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACADSB | NM_001609.3 | NP_001600.1 | ||
| XM_017016194.1 | XP_016871683.1 | |||
| AK298638.1 | ||||
| AK314241.1 | ||||
| AL831821.2 | CAD38535.2 | |||
| BC013756.1 | AAH13756.1 | |||
| DA964130.1 | ||||
| KU177879.1 | ||||
| KU177880.1 | ||||
| U12778.1 | AAA74424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map