Assay Details
Target Gene Details
Entrez Gene ID: | 8945 |
Gene Name: | beta-transducin repeat containing E3 ubiquitin protein ligase |
Gene Aliases: |
BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP |
Location: |
Chr.10:101353808-101557321 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTRC | NM_001256856.1 | NP_001243785.1 | ||
| NM_003939.4 | NP_003930.1 | |||
| NM_033637.3 | NP_378663.1 | |||
| XM_006718054.2 | XP_006718117.1 | |||
| XM_017016870.1 | XP_016872359.1 | |||
| XM_017016872.1 | XP_016872361.1 | |||
| XM_017016874.1 | XP_016872363.1 | |||
| AF101784.1 | AAD08702.1 | |||
| AF129530.1 | AAF04464.1 | |||
| AK295870.1 | ||||
| AK313353.1 | ||||
| AK313417.1 | ||||
| AL707686.1 | ||||
| AY207475.1 | AAO59171.1 | |||
| AY207476.1 | AAO59172.1 | |||
| BC027994.1 | AAH27994.1 | |||
| CR749295.1 | CAH18150.1 | |||
| DA218668.1 | ||||
| GU727631.1 | ||||
| KU178451.1 | ||||
| KU178452.1 | ||||
| KU178453.1 | ||||
| KU178454.1 | ||||
| Y14153.1 | CAA74572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv964n100 | Chr.10:101363048 - 101579801 on Build GRCh38 | Gain |
|
| esv2750847 | Chr.10:101295225 - 101692888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map