Assay Details
Target Gene Details
Entrez Gene ID: | 10659 |
Gene Name: | CUGBP, Elav-like family member 2 |
Gene Aliases: |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
Location: |
Chr.10:10462515-11336675 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF2 | NM_001326317.1 | NP_001313246.1 | ||
| NM_001326318.1 | NP_001313247.1 | |||
| NM_001326319.1 | NP_001313248.1 | |||
| NM_001326320.1 | NP_001313249.1 | |||
| NM_001326321.1 | NP_001313250.1 | |||
| NM_001326323.1 | NP_001313252.1 | |||
| NM_001326324.1 | NP_001313253.1 | |||
| NM_001326325.1 | NP_001313254.1 | |||
| NM_001326326.1 | NP_001313255.1 | |||
| NM_001326327.1 | NP_001313256.1 | |||
| NM_001326328.1 | NP_001313257.1 | |||
| XM_017015557.1 | XP_016871046.1 | |||
| XM_017015568.1 | XP_016871057.1 |
Target Gene Details
Entrez Gene ID: | 254312 |
Gene Name: | long intergenic non-protein coding RNA 710 |
Gene Aliases: |
- |
Location: |
Chr.10:10934940-10952163 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00710 | NR_015413.1 | |||
| NR_024410.1 | ||||
| NR_024411.1 | ||||
| DB038073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047445 | Chr.10:10789399 - 11147629 on Build GRCh38 | Gain |
|
| nsv549964 | Chr.10:10785719 - 10963741 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map