Assay Details
Target Gene Details
Entrez Gene ID: | 283050 |
Gene Name: | ZMIZ1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:78943326-79067448 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMIZ1-AS1 | NR_015429.1 | 6 | 566 | |
| NR_024429.1 | 6 | 662 | ||
| NR_024431.2 | 8 | 858 | ||
| AK097923.1 | 1 | 1066 | ||
| AK098249.1 | 8 | 1303 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467384 | Chr.10:78349364 - 79336832 on Build GRCh38 | Gain |
|
| nsv7343 | Chr.10:78955245 - 79019009 on Build GRCh38 | Insertion |
|

Genomic Map