Assay Details
Target Gene Details
Entrez Gene ID: | 84930 |
Gene Name: | microtubule associated serine/threonine kinase like |
Gene Aliases: |
GREATWALL, GW, GWL, MAST-L, THC2 |
Location: |
Chr.10:27154468-27187953 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASTL | NM_001172303.2 | NP_001165774.1 | ||
| NM_001172304.2 | NP_001165775.1 | |||
| NM_001320756.1 | NP_001307685.1 | |||
| NM_001320757.1 | NP_001307686.1 | |||
| NM_032844.4 | NP_116233.2 | |||
| NR_135469.1 | ||||
| XM_005252631.3 | XP_005252688.1 | |||
| XM_005252632.3 | XP_005252689.1 | |||
| XM_006717519.3 | XP_006717582.1 | |||
| XM_006717520.3 | XP_006717583.1 | |||
| XM_011519751.2 | XP_011518053.1 | |||
| XM_017016852.1 | XP_016872341.1 | |||
| XM_017016853.1 | XP_016872342.1 | |||
| XM_017016854.1 | XP_016872343.1 | |||
| XM_017016855.1 | XP_016872344.1 | |||
| XM_017016856.1 | XP_016872345.1 | |||
| AK027719.1 | BAB55321.1 | |||
| AK074804.1 | BAC11218.1 | |||
| AK123004.1 | ||||
| AK225319.1 | ||||
| BC009107.1 | AAH09107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622709 | Chr.10:27165209 - 27189102 on Build GRCh38 | Gain |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
|
| esv2422253 | Chr.10:27142437 - 27286441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map