Assay Details
Target Gene Details
Entrez Gene ID: | 5592 |
Gene Name: | protein kinase, cGMP-dependent, type I |
Gene Aliases: |
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha |
Location: |
Chr.10:50991151-52298350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKG1 | NM_001098512.2 | NP_001091982.1 | ||
| AK300110.1 | ||||
| D45864.1 | BAA08297.1 | |||
| DC310312.1 | ||||
| DC379897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831873 | Chr.10:50950850 - 51139780 on Build GRCh38 | Gain |
|
| nsv831872 | Chr.10:50903568 - 51058777 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79626168] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map