Assay Details
Target Gene Details
Entrez Gene ID: | 219736 |
Gene Name: | storkhead box 1 |
Gene Aliases: |
C10orf24 |
Location: |
Chr.10:68827537-68895942 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STOX1 | NM_001130159.2 | NP_001123631.1 | ||
| NM_001130160.2 | NP_001123632.1 | |||
| NM_001130161.2 | NP_001123633.1 | |||
| NM_152709.4 | NP_689922.3 | |||
| XM_011539454.2 | XP_011537756.1 | |||
| AK124681.1 | BAC85925.1 | |||
| AY842014.1 | AAW48524.1 | |||
| AY842015.1 | AAW48525.1 | |||
| AY842016.1 | AAW48526.1 | |||
| AY842017.1 | AAW48527.1 | |||
| BC140011.1 | ||||
| DA118241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437688 | Chr.10:68850453 - 68866528 on Build GRCh38 | Loss |
|
| dgv151e199 | Chr.10:68852253 - 68864038 on Build GRCh38 | Deletion |
|
| nsv1161974 | Chr.10:68851487 - 68866574 on Build GRCh38 | Duplication |
|
| nsv516725 | Chr.10:68856443 - 68859103 on Build GRCh38 | Loss |
|
| esv3623730 | Chr.10:68852320 - 68864024 on Build GRCh38 | Loss |
|
| nsv442191 | Chr.10:68852888 - 68862361 on Build GRCh38 | Loss |
|
| nsv1045343 | Chr.10:68853524 - 68866574 on Build GRCh38 | Loss |
|
| nsv8685 | Chr.10:68852817 - 68868800 on Build GRCh38 | Loss |
|
| esv25403 | Chr.10:68852829 - 68862542 on Build GRCh38 | Loss |
|
| esv2759764 | Chr.10:68697962 - 68871556 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map