Assay Details
Target Gene Details
Entrez Gene ID: | 196792 |
Gene Name: | family with sequence similarity 24 member B |
Gene Aliases: |
- |
Location: |
Chr.10:122849094-122879641 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM24B | NM_001204364.1 | NP_001191293.1 | ||
| NM_152644.2 | NP_689857.2 | |||
| NR_037911.1 | ||||
| AV762334.2 | ||||
| BC031343.1 | AAH31343.1 | |||
| BG490871.1 | ||||
| BU602099.1 | ||||
| BU958134.1 | ||||
| CB410280.1 |
Target Gene Details
Entrez Gene ID: | 100533195 |
Gene Name: | FAM24B-CUZD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.10:122832155-122879641 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM24B-CUZD1 | NR_037915.1 | |||
| AK300849.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|

Genomic Map