Assay Details
Target Gene Details
Entrez Gene ID: | 389941 |
Gene Name: | complement C1q like 3 |
Gene Aliases: |
C1QTNF13, C1ql, CTRP13, K100 |
Location: |
Chr.10:16513743-16522005 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QL3 | NM_001010908.1 | NP_001010908.1 | ||
| BC127716.1 | ||||
| BC127717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv25020 | Chr.10:16519159 - 16526059 on Build GRCh38 | Deletion |
|
| nsv831797 | Chr.10:16514510 - 16690309 on Build GRCh38 | Loss |
|
| nsv1046833 | Chr.10:16385108 - 16530259 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map