Assay Details
Target Gene Details
Entrez Gene ID: | 64072 |
Gene Name: | cadherin-related 23 |
Gene Aliases: |
CDHR23, USH1D |
Location: |
Chr.10:71396934-71815947 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Gene Details
Entrez Gene ID: | 102723377 |
Gene Name: | uncharacterized LOC102723377 |
Gene Aliases: |
- |
Location: |
Chr.10:71508153-71511920 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723377 | NR_120672.1 | |||
| BX103495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044971 | Chr.10:71272698 - 71673682 on Build GRCh38 | Gain |
|
| esv2759766 | Chr.10:71478146 - 71526398 on Build GRCh38 | Loss |
|
| nsv523021 | Chr.10:71480316 - 71585307 on Build GRCh38 | Gain |
|
| nsv551502 | Chr.10:71402480 - 71687524 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map