Assay Details
Target Gene Details
Entrez Gene ID: | 1763 |
Gene Name: | DNA replication helicase/nuclease 2 |
Gene Aliases: |
DNA2L, hDNA2 |
Location: |
Chr.10:68414064-68472521 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNA2 | NM_001080449.2 | 21 | 3700 | NP_001073918.2 |
| NR_102264.1 | 22 | 3565 | ||
| XM_006717680.3 | 22 | 4080 | XP_006717743.1 | |
| XM_011539417.1 | 15 | 2624 | XP_011537719.1 | |
| XM_017015799.1 | 18 | 3089 | XP_016871288.1 | |
| BC041115.2 | 9 | 1718 | AAH41115.1 | |
| BC053574.1 | 11 | 1894 | AAH53574.1 | |
| BC063664.1 | 17 | 2899 | AAH63664.1 | |
| D42046.1 | 21 | 3642 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv919n100 | Chr.10:68400084 - 68752800 on Build GRCh38 | Gain |
|
| nsv551434 | Chr.10:68384017 - 68503822 on Build GRCh38 | Loss |
|
| nsv1050943 | Chr.10:68414663 - 68511675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map