Assay Details
Target Gene Details
Entrez Gene ID: | 10771 |
Gene Name: | zinc finger MYND-type containing 11 |
Gene Aliases: |
BRAM1, BS69, MRD30 |
Location: |
Chr.10:134340-254637 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv651n100 | Chr.10:161632 - 739989 on Build GRCh38 | Gain |
|
| esv3622070 | Chr.10:118426 - 174375 on Build GRCh38 | Gain |
|
| dgv649n100 | Chr.10:26861 - 228735 on Build GRCh38 | Gain |
|
| nsv1037507 | Chr.10:97603 - 199464 on Build GRCh38 | Gain |
|
| dgv650n100 | Chr.10:83918 - 270899 on Build GRCh38 | Gain |
|
| esv2759723 | Chr.10:14061 - 178465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map