Assay Details
Target Gene Details
Entrez Gene ID: | 11164 |
Gene Name: | nudix hydrolase 5 |
Gene Aliases: |
YSA1, YSA1H, YSAH1 |
Location: |
Chr.10:12165328-12196144 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT5 | NM_001321647.1 | NP_001308576.1 | ||
| NM_001321648.1 | NP_001308577.1 | |||
| NM_014142.3 | NP_054861.2 | |||
| AF155832.1 | AAF06734.1 | |||
| AF161464.1 | AAF29079.1 | |||
| AF218818.1 | AAF25479.1 | |||
| AK291131.1 | ||||
| AK314144.1 | ||||
| BC000025.2 | AAH00025.1 | |||
| BU602000.1 | ||||
| CR457195.1 | CAG33476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035709 | Chr.10:12054410 - 12183299 on Build GRCh38 | Gain |
|
| nsv5820 | Chr.10:12155307 - 12187443 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map