Assay Details
Target Gene Details
Entrez Gene ID: | 56647 |
Gene Name: | BRCA2 and CDKN1A interacting protein |
Gene Aliases: |
TOK-1, TOK1 |
Location: |
Chr.10:125823535-125853695 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCCIP | NM_016567.3 | NP_057651.1 | ||
| NM_078468.2 | NP_510868.1 | |||
| NM_078469.2 | NP_510869.1 | |||
| AB040450.1 | BAA92927.1 | |||
| AB040451.1 | BAA92928.1 | |||
| AK055691.1 | ||||
| AK300767.1 | ||||
| AK304529.1 | ||||
| AL834458.1 | CAD39118.1 | |||
| AY064248.1 | AAL55439.1 | |||
| AY064249.1 | AAL55440.1 | |||
| AY092061.1 | ||||
| BC009771.1 | AAH09771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428241 | Chr.10:125765331 - 125928101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map