Assay Details
Target Gene Details
Entrez Gene ID: | 399806 |
Gene Name: | LBX1 antisense RNA 1 (head to head) |
Gene Aliases: |
MUSE |
Location: |
Chr.10:101229594-101238859 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LBX1-AS1 | NR_029380.1 | 3 | 699 | |
| AK096698.1 | 2 | 1940 | ||
| AK294034.1 | 3 | 592 | ||
| LC015092.1 | 3 | 717 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831966 | Chr.10:101124857 - 101287432 on Build GRCh38 | Gain |
|
| esv3624371 | Chr.10:101232878 - 101240073 on Build GRCh38 | Loss |
|

Genomic Map