Assay Details
Target Gene Details
Entrez Gene ID: | 643414 |
Gene Name: | lipase family member K |
Gene Aliases: |
LIPL2, bA186O14.2 |
Location: |
Chr.10:88706224-88753092 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPK | NM_001080518.1 | NP_001073987.1 | ||
| XM_011540073.2 | XP_011538375.1 | |||
| XM_011540075.1 | XP_011538377.1 | |||
| XM_011540077.1 | XP_011538379.1 | |||
| XM_017016545.1 | XP_016872034.1 | |||
| XM_017016546.1 | XP_016872035.1 | |||
| XM_017016547.1 | XP_016872036.1 | |||
| XM_017016548.1 | XP_016872037.1 | |||
| EF426482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551840 | Chr.10:88718726 - 88841108 on Build GRCh38 | Gain |
|
| nsv1043015 | Chr.10:88707310 - 88833935 on Build GRCh38 | Gain |
|
| nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map