Assay Details
Target Gene Details
Entrez Gene ID: | 23788 |
Gene Name: | mitochondrial carrier 2 |
Gene Aliases: |
HSPC032, MIMP, SLC25A50 |
Location: |
Chr.11:47604309-47642654 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTCH2 | XM_011519959.2 | XP_011518261.1 | ||
| XM_011519960.2 | XP_011518262.1 | |||
| XM_011519961.2 | XP_011518263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111949532,rs75092034] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map