Assay Details
Target Gene Details
Entrez Gene ID: | 51074 |
Gene Name: | APAF1 interacting protein |
Gene Aliases: |
APIP2, CGI-29, CGI29, MMRP19, hAPIP |
Location: |
Chr.11:34882295-34916411 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APIP | NM_015957.3 | NP_057041.2 | ||
| XM_011520154.2 | XP_011518456.1 | |||
| XM_017017875.1 | XP_016873364.1 | |||
| AF132963.1 | AAD27738.1 | |||
| AK225249.1 | ||||
| AK292648.1 | ||||
| AK302225.1 | ||||
| BC008440.1 | AAH08440.1 | |||
| BC009077.1 | AAH09077.1 | |||
| BC017594.2 | AAH17594.1 | |||
| BP200131.1 | ||||
| DA729016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8807 | Chr.11:34890620 - 34908742 on Build GRCh38 | Gain |
|
| esv2759817 | Chr.11:34891768 - 34942181 on Build GRCh38 | Loss |
|
| esv34511 | Chr.11:34892577 - 34921289 on Build GRCh38 | Loss |
|
| nsv832119 | Chr.11:34793981 - 34987823 on Build GRCh38 | Gain |
|
| esv3625916 | Chr.11:34891859 - 34899584 on Build GRCh38 | Loss |
|
| esv2666133 | Chr.11:34891880 - 34899608 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map