Assay Details
Target Gene Details
Entrez Gene ID: | 80162 |
Gene Name: | protein-glucosylgalactosylhydroxylysine glucosidase |
Gene Aliases: |
ATHL1 |
Location: |
Chr.11:287760-295688 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGGHG | XM_011520383.2 | XP_011518685.1 | ||
| XM_011520384.2 | XP_011518686.1 | |||
| XM_011520385.2 | XP_011518687.1 | |||
| XM_011520386.2 | XP_011518688.1 | |||
| XM_011520387.2 | XP_011518689.1 | |||
| XM_017018355.1 | XP_016873844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070914 | Chr.11:288899 - 289600 on Build GRCh38 | Deletion |
|
| esv3891905 | Chr.11:269147 - 336332 on Build GRCh38 | Gain |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1012n100 | Chr.11:198510 - 427836 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map