Assay Details
Target Gene Details
Entrez Gene ID: | 493900 |
Gene Name: | TMEM9B antisense RNA 1 |
Gene Aliases: |
C11orf18 |
Location: |
Chr.11:8964675-8976283 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM9B-AS1 | NR_073431.1 | |||
| AA339546.1 | ||||
| DA631400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv435923 | Chr.11:8817778 - 8979055 on Build GRCh38 | Deletion |
|
| esv2759803 | Chr.11:8763266 - 8988026 on Build GRCh38 | Gain |
|
| esv3891928 | Chr.11:8959990 - 9069697 on Build GRCh38 | Gain |
|
| nsv832062 | Chr.11:8827748 - 8987925 on Build GRCh38 | Loss |
|

Genomic Map