Assay Details
Target Gene Details
Entrez Gene ID: | 497258 |
Gene Name: | BDNF antisense RNA |
Gene Aliases: |
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049 |
Location: |
Chr.11:27506852-27698171 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BDNF-AS | NR_002832.2 | |||
| NR_033312.1 | ||||
| NR_033313.1 | ||||
| NR_033314.1 | ||||
| NR_033315.1 | ||||
| AY054391.4 | ||||
| AY054401.3 | ||||
| AY054402.3 | ||||
| AY054403.4 | ||||
| AY054404.4 | ||||
| AY054405.4 | ||||
| AY513486.3 | ||||
| EF689022.1 | ||||
| EF689023.1 | ||||
| EF689024.1 | ||||
| EF689025.1 | ||||
| EF689026.1 | ||||
| EF689027.1 | ||||
| EF689028.1 | ||||
| EF689029.1 | ||||
| EF689030.1 | ||||
| EF689031.1 | ||||
| EF689032.1 | ||||
| EF689033.1 | ||||
| EF689034.1 | ||||
| EF689035.1 | ||||
| EF689036.1 | ||||
| EF689037.1 | ||||
| EF689038.1 | ||||
| EF689039.1 | ||||
| EF689040.1 | ||||
| EF689041.1 | ||||
| EF689042.1 |
Target Gene Details
Entrez Gene ID: | 101410541 |
Gene Name: | long intergenic non-protein coding RNA 678 |
Gene Aliases: |
- |
Location: |
Chr.11:27617626-27634627 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00678 | NR_102708.1 | |||
| CD049147.1 | ||||
| CD049808.1 | ||||
| CD176172.1 | ||||
| EF565102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832095 | Chr.11:27517079 - 27694192 on Build GRCh38 | Gain |
|

Genomic Map