Assay Details
Target Gene Details
Entrez Gene ID: | 112937 |
Gene Name: | galactosidase beta 1 like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:134275447-134321364 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L3 | XM_017017155.1 | 1 | 851 | XP_016872644.1 |
| XM_017017156.1 | 1 | 851 | XP_016872645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1302n100 | Chr.11:134275705 - 134348093 on Build GRCh38 | Loss |
|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| nsv826148 | Chr.11:134273214 - 134282719 on Build GRCh38 | Gain |
|
| nsv468901 | Chr.11:134266299 - 134340635 on Build GRCh38 | Loss |
|
| nsv556598 | Chr.11:134252356 - 134298912 on Build GRCh38 | Loss |
|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
| nsv1037294 | Chr.11:134260280 - 134340956 on Build GRCh38 | Loss |
|
| nsv832299 | Chr.11:134170258 - 134346785 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139848733] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map