Assay Details
Target Gene Details
Entrez Gene ID: | 8050 |
Gene Name: | pyruvate dehydrogenase complex component X |
Gene Aliases: |
DLDBP, E3BP, OPDX, PDX1, proX |
Location: |
Chr.11:34915829-34996128 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDHX | NM_001135024.1 | NP_001128496.1 | ||
| NM_001166158.1 | NP_001159630.1 | |||
| NM_003477.2 | NP_003468.2 | |||
| XM_011520390.1 | XP_011518692.1 | |||
| AF001437.1 | AAB66315.1 | |||
| AK301384.1 | ||||
| AK312463.1 | ||||
| BC010389.1 | AAH10389.1 | |||
| BI915296.1 | ||||
| BX648910.1 | ||||
| DA573433.1 | ||||
| U82328.1 | AAC39661.1 | |||
| Y13145.1 | CAA73606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759817 | Chr.11:34891768 - 34942181 on Build GRCh38 | Loss |
|
| nsv437715 | Chr.11:34919947 - 34930446 on Build GRCh38 | Loss |
|
| esv34511 | Chr.11:34892577 - 34921289 on Build GRCh38 | Loss |
|
| nsv832119 | Chr.11:34793981 - 34987823 on Build GRCh38 | Gain |
|
| esv3579474 | Chr.11:34919435 - 34921446 on Build GRCh38 | Loss |
|
| nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map