Assay Details
Target Gene Details
Entrez Gene ID: | 10249 |
Gene Name: | glycine-N-acyltransferase |
Gene Aliases: |
ACGNAT, CAT, GAT |
Location: |
Chr.11:58708757-58731974 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYAT | NM_005838.3 | NP_005829.3 | ||
| NM_201648.2 | NP_964011.2 | |||
| XM_017017087.1 | XP_016872576.1 | |||
| AB013093.1 | ||||
| AB665285.1 | ||||
| AF023466.1 | AAB81453.1 | |||
| BC009785.1 | AAH09785.1 | |||
| BG401676.1 | ||||
| BG617408.1 | ||||
| BP277146.1 | ||||
| CB161978.1 | ||||
| CR456927.1 | CAG33208.1 | |||
| DR004166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832168 | Chr.11:58626587 - 58797178 on Build GRCh38 | Gain |
|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map