Assay Details
Target Gene Details
Entrez Gene ID: | 6392 |
Gene Name: | succinate dehydrogenase complex subunit D |
Gene Aliases: |
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS |
Location: |
Chr.11:112086824-112095801 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHD | NM_001276503.1 | NP_001263432.1 | ||
| NM_001276504.1 | NP_001263433.1 | |||
| NM_001276506.1 | NP_001263435.1 | |||
| NM_003002.3 | NP_002993.1 | |||
| NR_077060.1 | ||||
| AB006202.1 | BAA22054.1 | |||
| AK075360.1 | ||||
| BC005263.1 | AAH05263.1 | |||
| BC009574.1 | AAH09574.1 | |||
| BC012603.1 | AAH12603.1 | |||
| BC015188.1 | AAH15188.1 | |||
| BC015992.1 | AAH15992.1 | |||
| BC022350.1 | AAH22350.1 | |||
| BC070307.1 | AAH70307.1 | |||
| BC071755.1 | AAH71755.1 | |||
| BC071756.1 | AAH71756.1 | |||
| BE567994.1 | ||||
| BF211358.1 | ||||
| BG492859.1 | ||||
| BQ932668.1 | ||||
| BT007238.1 | AAP35902.1 | |||
| BX419391.2 | ||||
| CD105572.1 | ||||
| CR456932.1 | CAG33213.1 | |||
| KU178325.1 | ||||
| KU178326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map