Assay Details
Target Gene Details
Entrez Gene ID: | 92292 |
Gene Name: | glycine-N-acyltransferase like 1 |
Gene Aliases: |
GATF-C, GNAT |
Location: |
Chr.11:58927629-58957074 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYATL1 | NM_001220494.2 | NP_001207423.1 | ||
| NM_001220496.2 | NP_001207425.1 | |||
| NM_080661.4 | NP_542392.2 | |||
| XM_011545354.2 | XP_011543656.1 | |||
| XM_011545355.2 | XP_011543657.1 | |||
| XM_011545356.2 | XP_011543658.1 | |||
| XM_011545357.2 | XP_011543659.1 | |||
| XM_011545358.2 | XP_011543660.1 | |||
| XM_011545359.2 | XP_011543661.1 | |||
| AF087878.1 | AAP97178.1 | |||
| AK055818.1 | BAB71023.1 | |||
| AK091965.1 | BAC03779.1 | |||
| AK300479.1 | ||||
| BC008353.1 | AAH08353.1 | |||
| DC339353.1 | ||||
| DQ084381.1 |
Target Gene Details
Entrez Gene ID: | 283194 |
Gene Name: | uncharacterized LOC283194 |
Gene Aliases: |
- |
Location: |
Chr.11:58933643-59058452 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC283194 | NR_033853.2 | |||
| AK091446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv14n68 | Chr.11:58932624 - 59111366 on Build GRCh38 | Gain |
|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
| dgv139e201 | Chr.11:58845738 - 59113363 on Build GRCh38 | Deletion |
|
| nsv482929 | Chr.11:58932626 - 59094208 on Build GRCh38 | Gain |
|
| nsv428258 | Chr.11:58932619 - 59111366 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map