Assay Details
Target Gene Details
Entrez Gene ID: | 4723 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit V1 |
Gene Aliases: |
CI-51K, CI51KD, UQOR1 |
Location: |
Chr.11:67606852-67612541 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV1 | NM_001166102.1 | NP_001159574.1 | ||
| NM_007103.3 | NP_009034.2 | |||
| AF053070.1 | AAC39722.1 | |||
| AF092131.1 | AAD40373.1 | |||
| AK055875.1 | ||||
| AK223061.1 | BAD96781.1 | |||
| AK293522.1 | ||||
| AK300726.1 | ||||
| AK316122.1 | ||||
| BC007619.1 | AAH07619.1 | |||
| BC008146.1 | AAH08146.1 | |||
| BC015645.2 | AAH15645.1 | |||
| CR456739.1 | CAG33020.1 | |||
| DB532599.1 | ||||
| S67973.1 | AAB29698.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1981n54 | Chr.11:67579272 - 68049469 on Build GRCh38 | Gain |
|
| esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication |
|
| dgv154n27 | Chr.11:67605006 - 67666398 on Build GRCh38 | Loss |
|
| dgv1214n100 | Chr.11:67552445 - 67996597 on Build GRCh38 | Gain |
|
| nsv951023 | Chr.11:67608030 - 67649929 on Build GRCh38 | Deletion |
|
| nsv468606 | Chr.11:67585218 - 68049469 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112015076] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map