Assay Details
Target Gene Details
Entrez Gene ID: | 23193 |
Gene Name: | glucosidase II alpha subunit |
Gene Aliases: |
G2AN, GIIA, GLUII |
Location: |
Chr.11:62624826-62646726 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GANAB | NM_001278192.1 | NP_001265121.1 | ||
| NM_001278193.1 | NP_001265122.1 | |||
| NM_001278194.1 | NP_001265123.1 | |||
| NM_198334.2 | NP_938148.1 | |||
| NM_198335.3 | NP_938149.2 | |||
| XM_017017412.1 | XP_016872901.1 | |||
| XM_017017413.1 | XP_016872902.1 | |||
| XM_017017414.1 | XP_016872903.1 | |||
| AF144074.1 | AAF66685.1 | |||
| AJ000332.1 | CAA04006.1 | |||
| AK026548.1 | ||||
| AK026997.1 | ||||
| AK295806.1 | ||||
| AK299819.1 | ||||
| AK302752.1 | ||||
| AK314370.1 | ||||
| BC017433.2 | AAH17433.2 | |||
| BC017435.2 | AAH17435.2 | |||
| BC065266.1 | AAH65266.1 | |||
| D42041.1 | BAA07642.1 | |||
| EU794613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain |
|
| nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain |
|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map