Assay Details
Target Gene Details
Entrez Gene ID: | 390226 |
Gene Name: | guanylate cyclase 2E, pseudogene |
Gene Aliases: |
GC-E, GCD, GUCY2E |
Location: |
Chr.11:76680166-76721789 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY2EP | NR_024042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509433 | Chr.11:76670774 - 76728370 on Build GRCh38 | Insertion |
|
| nsv1047069 | Chr.11:76656642 - 76709769 on Build GRCh38 | Gain |
|
| dgv1228n100 | Chr.11:76590272 - 76729177 on Build GRCh38 | Loss |
|

Genomic Map