Assay Details
Target Gene Details
Entrez Gene ID: | 219970 |
Gene Name: | glycine-N-acyltransferase like 2 |
Gene Aliases: |
BXMAS2-10, GATF-B |
Location: |
Chr.11:58834065-58905699 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYATL2 | NM_145016.3 | NP_659453.3 | ||
| XM_011544809.2 | XP_011543111.1 | |||
| XM_017017337.1 | XP_016872826.1 | |||
| XM_017017338.1 | XP_016872827.1 | |||
| AB207211.1 | ||||
| AF426250.1 | AAO73139.1 | |||
| BC016789.1 | AAH16789.1 | |||
| BC021682.1 | AAH21682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
| dgv71n21 | Chr.11:58786298 - 58864895 on Build GRCh38 | Loss |
|
| esv3626524 | Chr.11:58832286 - 58836413 on Build GRCh38 | Loss |
|
| dgv1207n100 | Chr.11:58767187 - 58911023 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111680745] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map